missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies CryoStor™ CSB
Animal component-free and protein-free cryopreservation formulation
£737.10 - £7443.90
Specifications
Cell Type | B Cells, CHO Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells |
---|---|
For Use With (Application) | Cryopreservation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17137501
|
STEMCELL Technologies
100-0237 |
100 mL |
£737.10
100mL |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17147501
|
STEMCELL Technologies
100-0238 |
500 mL |
£3692.25
500mL |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17157501
|
STEMCELL Technologies
100-0239 |
1000 mL |
£7443.90
1000mL |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- CryoStor™ CSB is an intracellular-like base formulation from the CryoStor™ preservation media series
- It is serum-free, animal component-free, and does not contain dimethyl sulfoxide (DMSO)
- It is designed to improve post-thaw cell viability and function, and to reduce the level of cryopreservation-induced delayed-onset cell death
Specifications
B Cells, CHO Cells, Hybridomas, Intestinal Cells, Macrophages, Mesenchymal Stem and Progenitor Cells, Monocytes, Myeloma, NK Cells, Other, Pluripotent Stem Cells, T Cells | |
Cryopreservation |