missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Graduated Polypropylene Cryotubes with stand

Polypropylene with highest translucence. Use these tubes for the storing and conservation of tissue cultures and biological samples. Fisherbrand™ Graduated Polypropylene Cryotubes are ideal for applications to -90°C in the gaseous phase of liquid nitrogen or in freezers.

Brand:  Fisherbrand™ CRY-260-030A

Product Code. 10083442

  • £586.00 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Description

Description

  • Sterilized by beta-radiation according to EN ISO 11137
  • Lid with integrated sealing function; no additional silicone O-ring
  • Internal conical bottom for complete emptying
  • White graduations for precise measurement
  • White lettering for sample identification
  • Available with internal or external threads
  • Equipped with base for using with one hand
  • 2.0 ml, 4.0 ml and 5.0 ml tubes available also with round bottom
  • Packaged in PE bags of 50 tubes in boxes
  • Free of DNA, DNase, pyrogens and ATP
Specifications

Specifications

Natural
2 mL
Stand
12.5 mm
Polypropylene
49 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Graduated Polypropylene Cryotubes with stand >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.