missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Tygon™ LMT-55 Tubing without Configuration
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Tygon™ LMT-55 Tubing without Configuration handles virtually all inorganic chemicals found in the lab.
£58.75 - £79.20
Specifications
Product Type | Tubing |
---|---|
Material | Tygon R3607 |
Overall Length (English) | 32.8 ft. |
Overall Length (Metric) | 10 m |
For Use With (Application) | For sample and reagent dispensing, biological and chemical assays, and water and soil analysis |
Product Code | Brand | Color | Diameter (Metric) Inner | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (Metric) Inner | Price | Quantity & Availability | ||||
15113226
|
Saint Gobain Life Sciences
070534-07L-ND |
Orange/White | 0.64 mm |
£79.20
10m |
Estimated Shipment: 25-10-2024
Log in to see stock. |
||||
15153226
|
Saint Gobain Life Sciences
070534-10L-ND |
White/Black | 0.95 mm |
£71.25
10m |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||||
15183226
|
Saint Gobain Life Sciences
070534-13L-ND |
Red/Red | 1.14 mm |
£66.55
10m |
Estimated Shipment: 21-10-2024
Log in to see stock. |
||||
10092393
|
Saint Gobain Life Sciences
070534-18L-ND |
Blue/Blue | 1.65 mm |
£64.65
10m |
Estimated Shipment: 28-11-2024
Log in to see stock. |
||||
15183216
|
Saint Gobain Life Sciences
070534-05L-ND |
Orange/Yellow | 0.51 mm |
£66.55
10m |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15123226
|
Saint Gobain Life Sciences
070534-09L-ND |
Crystal Clear | 0.89 mm |
£58.75
10m |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||||
15133236
|
Saint Gobain Life Sciences
070534-21L-ND |
Purple/Purple | 2.06 mm |
£58.75
10m |
Estimated Shipment: 04-11-2024
Log in to see stock. |
||||
15133226
|
Saint Gobain Life Sciences
070534-11L-ND |
Crystal Clear | 1.02 mm |
£58.75
10m |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
12068908
|
Saint Gobain Life Sciences
070534-24L-ND |
Purple/White | 2.79 mm |
£58.75
10m |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||||
Description
- Bio-based formulation delivers excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
- Resistant to flex fatigue and abrasion
- Less permeable than rubber tubing
- For use with virtually all inorganic chemicals found in the lab
Specifications
Tubing | |
32.8 ft. | |
For sample and reagent dispensing, biological and chemical assays, and water and soil analysis | |
-50°C | |
165.2°F |
Tygon R3607 | |
10 m | |
-58°F | |
1 Ea. | |
74°C |