Learn More
Multiroir™ Crystal Polystyrene Transmission Lab Box
Use these boxes for a variety of storage needs. Multiroir™ Crystal Polystyrene Transmission Lab Boxes are ideal for conservation, storage and presentation of products. Boxes are made of crystal polystyrene, a resistant material obtained by polymerization of styrene monomer CH2 = CH-Ph.
Brand:  Multiroir™ 45106
 View more versions of this product
Product Code. 11762944
  • £101.00 / Each
Explore more special offers
Description
Description
  • Transparent, stackable and highly resistant to UV
  • Integrated Label holder
  • Chemically inert plastic
  • Food contact quality
  • Funds and above have male-female reliefs that lock stacking and provide perfect storage stability
  • The tight tolerances of the fitting of the cover on the base ensure a good seal
Specifications
Specifications
10.85 L
Polystyrene
Conservation, Storage and Presentation of Products
1/Pk.
360 mm
240 mm
130 mm
Transparent
Rectangular
Transmission Lab Box
360 L x 138 H mm
355 mm
140 mm
0.96 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Multiroir™ Crystal Polystyrene Transmission Lab Box > Capacity: 10.85L; Dimensions: 360L x 138mmH