missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Crystal Violet Product Code.: 11435027

Thermo Scientific Chemicals Crystal Violet

Product Code. 11435027
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11435027

Brand: Thermo Scientific Chemicals B21932.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crystal Violet is used as an acid-base indicator, alcohol denaturant, biological stain, textile dye, and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.

Solubility
Miscible with water and chloroform. Partially miscible with alcohol and glycerol.

Notes
Incompatible with acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-62-9
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
MDL Number MFCD00011750
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
Synonym Basic Violet 3; C.I. 42555
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Spécification

Melting Point ∼215°C (decomposition)
UN Number UN3077
Beilstein 4077708
Merck Index 14,4395
Solubility Information Miscible with water and chloroform. Partially miscible with alcohol and glycerol.
Formula Weight 407.99
Quantity 25 g
Chemical Name or Material Crystal Violet

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250274 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberB21932.14,B21932.36,B21932.22
Lot Number10250370 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberB21932.14,B21932.36,B21932.22
Lot Number10247072 Certificate TypeCertificate of Analysis Date20/06/2023 Catalog NumberB21932,B21932.14,B21932.36,B21932.22
Lot Number10242917 Certificate TypeCertificate of Analysis Date17/08/2022 Catalog NumberB21932.14,B21932.36,B21932.22
Lot Number10237336 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB21932.14,B21932.36,B21932.22
compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Crystal Violet
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
Mention de danger
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contains : C.I. Basic violet 1

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit
Thermo Scientific Chemicals Crystal Violet >

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis