Learn More
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet and orange
Brand:  Cole-Parmer™ 45520-33
 View more versions of this product
Product Code. 15345046
  • £37.15 / Pack of 25
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
  • Superior chemical resistance compared to polycarbonate.
  • Sterilize by autoclave, gamma irradiation or ethylene oxide.
  • Meets USP Class VI.
  • Animal-derived ingredient-free.
  • Available in packs of 25.
Specifications
Specifications
Male luer lock
25/Pk.
Male Luer Lock Fitting
Polyolefin
Lime
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb > Lime