missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet and orange
£30.20 - £48.90
Specifications
Description | Male luer lock |
---|---|
Material | Polyolefin |
Packaging Quantity | 25/Pk. |
Product Type | Male Luer Lock Fitting |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
15305046
|
Cole-Parmer™
45520-03 |
Berry |
£30.20
Pack of 25 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
15315046
|
Cole-Parmer™
45520-13 |
Rose |
£30.20
Pack of 25 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
15325046
|
Cole-Parmer™
45520-23 |
Yellow |
£30.20
Pack of 25 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
15345046
|
Cole-Parmer™
45520-33 |
Lime |
£37.15
Pack of 25 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
15355046
|
Cole-Parmer™
45520-53 |
Orange |
£48.90
Pack of 25 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
Description
- Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
- Superior chemical resistance compared to polycarbonate.
- Sterilize by autoclave, gamma irradiation or ethylene oxide.
- Meets USP Class VI.
- Animal-derived ingredient-free.
- Available in packs of 25.
Specifications
Male luer lock | |
Polyolefin | |
25/Pk. | |
Male Luer Lock Fitting |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb