Learn More
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet and orange
Brand:  Cole-Parmer™ 45520-53
 View more versions of this product
Product Code. 15355046
  • £48.90 / Pack of 25
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
  • Superior chemical resistance compared to polycarbonate.
  • Sterilize by autoclave, gamma irradiation or ethylene oxide.
  • Meets USP Class VI.
  • Animal-derived ingredient-free.
  • Available in packs of 25.
Specifications
Specifications
Male luer lock
25/Pk.
Male Luer Lock Fitting
Polyolefin
Orange
Product Suggestions
Show More
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ CrystalVu™ Luer Fitting with Male Lueur Lock x 1/8 in. Hose Barb > Orange