Learn More
BD Plastipak™ Plastic Concentric Luer-Lock Syringe
Enables accurate dosing and visualisation of the syringe contents. BD Medical™ Plastipak™ Plastic Concentric Luer-Lock Syringe are each tested for sterility, pyrogenicity and toxicity.
Brand:  BD 301229
 View more versions of this product
Product Code. 10109922
  • £395.00 / Pack of 60
Edge
Explore more special offers
Description
Description
  • Concentric Luer-Lok
  • Latex free stopper
  • CE marked
  • Polypropylene
  • Sterile
  • Disposable
  • Without needleFor needles, please see our Microlance™ disposable needles range. All products are for research use only, unless otherwise indicated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Mark, European Directive 93/42 / EEC (compliance procedure Annex V and VII) Certificates: No. 95 Q 252156 and No. 252231 Q (references 309661 & 309620 ) ; certified by NSAI (No. 0050)
Transparent
For use with automatic pumps and they are suitable for most the pumps
107.5 mm
Single-use
Pk. of 60
Concentric Luer Lock
Sterile
Plastic
23.77 mm
Yes
Concentric Luer-lock Syringe
1 mL
30 mL
Yes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Plastipak™ Plastic Concentric Luer-Lock Syringe > Outside Diameter: 23.77mm; Volume: 30mL