Learn More
BD Biosciences™ CS&T Research Beads for use with BD FACSDiva™ Version 7 or Later (RUO)
Automatically characterize, track, and report measurements of supported BD digital flow cytometers.
Brand:  BD Biosciences™ 655051
291.51 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15846329
  • £682.00 / Each
Explore more special offers
Description
Description
  • Designed for use on BD™ flow cytometers running BD FACSuite™ software version 7.0 or later.
  • FC beads enable the software to automatically characterize, track, and report measurements of supported BD digital flow cytometers.
  • CS&T research beads are dyed with fluorochromes that are excited by the cytometer's lasers.
  • Provides cost efficiency with low per-test cost and multiple kit configurations.
  • Produced under GMP for improved quality and reproducibility.
  • Enhances the cytometer setup workflow, supporting over 100 laser and filter combinations.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Three 3 mL vial at 50 tests per vial for 150 tests per kit
150 Tests
The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 24 hours at 2°C–8°CC or 8 hours at 15°C–25°C when protected from light
Fluorescent Polystyrene Beads
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Biosciences™ CS&T Research Beads for use with BD FACSDiva™ Version 7 or Later (RUO) > 150 Tests