missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-HIS, Powdered Media
Single dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media
Brand: MP Biomedicals™ 114510322
This item is not returnable.
View return policy
Description
Formulation
- Add 0.77g of CSM-His to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes
Applications:
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Molecular biology Certified™ | |
Powder | |
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L | |
100 g |
15°C to 30°C | |
Yeast Media | |
For growth of Saccharomyces cerevisiae |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction