missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-HIS, Powdered Media

Single dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

£57.60 - £504.00

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11377889
View Documents
MP Biomedicals™
114510312
10 g
£57.60
10g
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
11387889
View Documents
MP Biomedicals™
114510322
100 g
£504.00
100g
Estimated Shipment: 21-02-2025
to see stock.
Add to basket
 
Description

Description

Formulation

  • Add 0.77g of CSM-His to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes

Applications:

Yeast cell culture, Gene expression, Transcription regulation

Specifications

Specifications

Molecular biology Certified™
Powder
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 20mg/L, L-Valine 140mg/L
15°C to 30°C
Yeast Media
For growth of Saccharomyces cerevisiae
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 4 Years

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.