missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ CSM-URA, Powdered Media

Single dropout of Complete Supplement Mixture (CSM) of amino acids for S. cereviseae growth media

Brand:  MP Biomedicals™ 114511212

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11377899

  • £87.77 / 10g
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

  • Add 0.77g of CSM-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined single drop-out medium for S.cerevisiae

Applications:

Yeast cell culture, Gene expression, Transcription regulation

Specifications

Specifications

Molecular biology Certified™
Powder
Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 20mg/L, L-Isoleucine 50mg/L, L-Leucine 100mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L
10 g
15°C to 30°C
Yeast Media
For growth of Saccharomyces cerevisiae
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers