Learn More
Gibco™ CTS™ HiFi Cas9 Protein
Gibco CTS HiFi Cas9 Protein is our GMP-manufactured high-fidelity CRISPR-Cas9 protein, engineered to deliver reduced off-target effects while maintaining high on-target activity.
£12563.00 - £24066.00
Specifications
Concentration | 10 mg/mL |
---|---|
Product Type | Cas9 Protein |
Content And Storage | Store at –20°C. |
Shipping Condition | Dry Ice |
Expression System | E. coli |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17954721
|
Gibco™
A54223 |
2.5 mg |
£12563.00
2.50mg |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17974661
|
Gibco™
A54224 |
5 mg |
£24066.00
5mg |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Gibco CTS HiFi Cas9 Protein is our GMP-manufactured high-fidelity CRISPR-Cas9 protein, engineered to deliver reduced off-target effects while maintaining high on-target activity. This protein is designed and manufactured in compliance with the principles of GMP and to meet requirements of the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products. It is provided in a high-concentration and large-scale format for process development and clinical research applications.
Features of CTS HiFi Cas9 Protein include:
• Exceptional off-target profile—over 70% reduction in off-target effects in primary immune cells, compared to CTS TrueCut Cas9 (wild type) at tested loci
• Maintain on-target activity—achieve comparable on-target activity to CTS TrueCut Cas9 (wild type) in primary immune cells at tested loci
• Stringent manufacturing process—manufactured to meet the standards for Ancillary Materials for Cell, Gene, and Tissue-Based Products including USP , Ph.Eur. 5.2.12, and ISO 20399 following the principles of 21 CFR Part 820 in an FDA-registered manufacturing site
• Extensive safety testing—includes sterility, endotoxin, mycoplasma, residual nucleic acid, and protein testing
• Traceability documentation—COA, COO, Drug Master files, and Regulatory Support files* are available
CTS HiFi Cas9 Protein is a Streptococcus pyogenes high-fidelity Cas9 protein, produced by recombinant DNA technology in E. coli for genome editing with CRISPR technology. Cas9 protein forms a very stable RNP complex with the guide RNA (gRNA) component of the CRISPR-Cas9 system. Incorporation of NLS aids delivery to the nucleus, increasing the editing efficiency and on-target cleavage.
If you would like to evaluate our HiFi Cas9 protein or conduct small-scale research experiments, we recommend using our research-grade TrueCut HiFi Cas9 Protein (Cat. No. A50576).
*Country-specific Regulatory Support File process will be initiated upon request and will require additional documentation time.
Specifications
10 mg/mL | |
Store at –20°C. | |
E. coli | |
Genome Editing |
Cas9 Protein | |
Dry Ice | |
Liquid | |
ISO 13485 |