missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Cubis™ II Precision (3-place) Balance, MCA User Interface
GREENER_CHOICEBIOPHARMA

With completely configurable hardware, software, and connectivity, Cubis™ II offers a high-performance balance that will align with your unique demands and compliance requirements.

£7040.00 - £14980.00

Specifications

Certifications/Compliance 21 CFR PART 11 17025 USP 41
Connection Type 2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Display Type Color TFT Touchscreen
Leveling Automation motorized
Material Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
View More Specs

Products 6
Product Code Brand Capacity Includes Model Price Quantity & Availability  
Product Code Brand Capacity Includes Model Price Quantity & Availability  
16406945
BIOPHARMA
View Documents Promotion Details
Sartorius
MCA623S-2S00-E
620 g Balance, Accessories, Power supply, Manual, Factory test report Draft shield E: Small glass
£7040.00
Each
Please to purchase this item. Need a web account? Register with us today!
16223242
GREENER_CHOICEBIOPHARMA
View Documents
Sartorius
MCA623S-2S00-E QP99
620 g Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield E: Small glass
£9225.00
1 unit
Please to purchase this item. Need a web account? Register with us today!
16203182
BIOPHARMA
View Documents
Sartorius
MCA1203S-2S00-E QP99
1.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield E: Small glass
£11565.00
1 unit
Please to purchase this item. Need a web account? Register with us today!
16203202
BIOPHARMA
View Documents
Sartorius
MCA2203S-2S00-E QP99
2.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield: E: Small glass
£12290.00
1 unit
Please to purchase this item. Need a web account? Register with us today!
16243212
GREENER_CHOICEBIOPHARMA
View Documents
Sartorius
MCA3203S-2S00-E QP99
3.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield E: Small glass
£12580.00
1 unit
Please to purchase this item. Need a web account? Register with us today!
16283222
BIOPHARMA
View Documents
Sartorius
MCA5203S-2S00-E QP99
5.2 kg Balance, Accessories, Power supply, Manual, Factory test report, QP99 software Draft shield E: Small glass
£14980.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • 21 CFR Part 11 compliance
  • Integrated audit-trail
  • State-of-the-art user management
  • Oustanding service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • For all models up to a maximum capacity of 8.2 kg
  • Large 7-inch color touch TFT display in 16:9 format with new user interface
  • Software Factory installed basic set of essential weighing applications (license free) and packages with special weighing applications and function extensions (license required)
  • Operation Activated by touch key, touch-free using IR sensor

    (draft shield M) or gesture sensor (optional), learning capability

Warranty

10-year warranty

Specifications

Specifications

21 CFR PART 11 17025 USP 41
Color TFT Touchscreen
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
Precision Balance
240 mm
120/240 V
Class I
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Automation motorized
0°C to +40°C
10 years
122/284/373 mm
1.0 A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Cubis™ II Precision (3-place) Balance, MCA User Interface

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.