Learn More
Sartorius Cubis™ II Precision Lab Balance
Maximum load between 320 g and 14,200 g and a readability of 1 mg to 100 mg provide, ideal model for every application.
£4880.00 - £13575.00
Specifications
Certifications/Compliance | Balance with Type Approval Certificate for China |
---|---|
Connection Type | D-sub connector, 9-pin |
Display Type | Color Touchscreen |
Material | Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim |
Electrical Requirements | 100 to 240 VAC (± 10%), 50/60 Hz, 1.0 A |
Product Code | Brand | Capacity | Readability | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Model | Price | Quantity & Availability | |||
16213252
![]() |
Sartorius
MCE12201S0-CEU |
12,200 g | 100 mg | Weighing pan without draft shield |
£5625.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16293252
![]() |
Sartorius
MCE2202S0-CEU |
2200 g | 10 mg | Weighing pan without draft shield |
£5135.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16213272
![]() |
Sartorius
MCE32202P0-CEU |
4200 g, 32200 g | 10 mg, 100 mg | Weighing pan without draft shield |
£13575.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16293272
![]() |
Sartorius
MCE4202S0-CEU |
14,200 g | 10 mg | Weighing pan without draft shield |
£5440.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16213282
![]() |
Sartorius
MCE6202S0-CEU |
6200 g | 10 mg | Weighing pan without draft shield |
£6320.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16253282
![]() |
Sartorius
MCE8201S0-CEU |
8200 g | 100 mg | Weighing pan without draft shield |
£4880.00
1 unit |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16467195
![]() |
Sartorius
MCE1203S-2CEU-R |
1200 g | 1 mg | Draft Shield R: Flat, stainless steel weighing pan draft shield (removable, with no glass components) for all precision balances |
£8940.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16477195
![]() |
Sartorius
MCE1203S-2CEU-U |
1200 g | 1 mg | Draft shield U: Manual glass |
£9775.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16437255
![]() |
Sartorius
MCE2203P-2CEU-E |
1010 g, 2200 g | 1 mg | Draft Shield E: Manual glass draft shield for precision balances |
£8855.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16407275
![]() |
Sartorius
MCE2203S-2CEU-R |
2200 g | 1 mg | Draft Shield R: Flat, stainless steel weighing pan draft shield (removable, with no glass components) for all precision balances |
£9660.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16457315
![]() |
Sartorius
MCE3203S-2CEU-R |
3200 g | 1 mg | Draft Shield R: Flat, stainless steel weighing pan draft shield (removable, with no glass components) for all precision balances |
£9955.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16467465
![]() |
Sartorius
MCE623S-2CEU-E |
620 g | 1 mg | Draft Shield E: Manual glass draft shield for precision balances |
£6595.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
16487465
![]() |
Sartorius
MCE623S-2CEU-R |
620 g | 1 mg | Draft Shield R: Flat, stainless steel weighing pan draft shield (removable, with no glass components) for all precision balances |
£6595.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifications
Balance with Type Approval Certificate for China | |
Color Touchscreen | |
100 to 240 VAC (± 10%), 50/60 Hz, 1.0 A |
D-sub connector, 9-pin | |
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim |