Learn More
Sartorius Cubis™ II Precision Lab Balance
The Cubis™ II Precision Lab Balance with a maximum load between 320 g and 14,200 g and a readability of 1 mg to 100 mg provides an ideal model for every application.
£5450.00 - £15420.00
Specifications
Certifications/Compliance | Balance with Type Approval Certificate for China |
---|---|
Connection Type | D-sub connector, 9-pin |
Display Type | Color Touchscreen |
Material | Aluminum, plastic PBT, Optiwhite float glass and stainless steel |
Electrical Requirements | 100 – 240 VAC (±10%), 50 - 60 Hz, 1.0 A |
Product Code | Brand | Capacity | Readability | Includes | Model | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Readability | Includes | Model | Price | Quantity & Availability | ||
16424705
|
Sartorius
MCA323S-2CEU-E QP99 |
320 g | 1 mg | Draft Shield E, QP99 | CEU Electronic Toploading Balance |
£8265.00
Each |
Estimated Shipment: 16-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16213242
|
Sartorius
MCA623S-2CEU-E QP99 |
620 g | 1 mg | Draft Shield E, QP99 | CEU Electronic Toploading Balance |
£9085.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16233242
|
Sartorius
MCA623S-2CEU-R QP99 |
620 g | 1 mg | Draft Shield R, QP99 | CEU Electronic Toploading Balance |
£9085.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16293172
|
Sartorius
MCA1203S-2CEU-E QP99 |
1200 g | 1 mg | Draft Shield E, QP99 | CEU Electronic Toploading Balance |
£11445.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16213182
|
Sartorius
MCA1203S-2CEU-R QP99 |
1200 g | 1 mg | Draft Shield R,QP99 | CEU Electronic Toploading Balance |
£11445.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16466465
|
Sartorius
MCA6202P-2CEU-0 |
6200 g | 10, 20, 50 mg | Draft Shield O | CEU Electronic Toploading Balance |
£5450.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16293192
|
Sartorius
MCA2203S-2CEU-E QP99 |
2200 g | 1 mg | Draft Shield E, QP99 | CEU Electronic Toploading Balance |
£11995.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16213202
|
Sartorius
MCA2203S-2CEU-R QP99 |
2200 g | 1 mg | Draft Shield F, Draft Shield R, QP99 | CEU Electronic Toploading Balance |
£11995.00
1 unit |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16273192
|
Sartorius
MCA2202S-2CEU-0 QP99 |
2200 g | 10 mg | Draft Shield O, QP99 | CEU Electronic Toploading Balance |
£7720.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16233212
|
Sartorius
MCA3203S-2CEU-E QP99 |
3200 g | 1 mg | Draft Shield E, QP99 | CEU Electronic Toploading Balance |
£12275.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16253212
|
Sartorius
MCA3203S-2CEU-R QP99 |
3200 g | 1 mg | Draft Shield R, QP99 | CEU Electronic Toploading Balance |
£12275.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16463285
|
Sartorius
MCA14202S-2CEU-0 QP99 |
14200 g | 10 mg | Draft Shield O | CEU Electronic Toploading Balance |
£12870.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16253222
|
Sartorius
MCA4202S-2CEU-0 QP99 |
4200 g | 10 mg | Draft Shield O | CEU Electronic Toploading Balance |
£8000.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16273222
|
Sartorius
MCA5203S-2CEU-E QP99 |
5200 g | 1 mg | Draft Shield E, QP99 | CEU Electronic Toploading Balance |
£14545.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16475925
|
Sartorius
MCA5203S-2CEU-R |
5200 g | 1 mg | Draft Shield R | CEU Electronic Toploading Balance |
£12425.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16293222
|
Sartorius
MCA5203S-2CEU-R QP99 |
5200 g | 1 mg | Draft Shield R,QP99 | CEU Electronic Toploading Balance |
£14545.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16293232
|
Sartorius
MCA6202S-2CEU-0 QP99 |
6200 g | 10 mg | Draft Shield O | CEU Electronic Toploading Balance |
£8830.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16447165
|
Sartorius
MCA8202S-2S00-0 QP1 |
8200 g | 10 mg | Draft Shield O, QP1 | Standard Electronic Toploading Balance |
£8250.00
Each |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16342046
|
Sartorius
MCA8202S-2S00-0 |
8200 g | 10 mg | Draft Shield O, QP99 | Standard Electronic Toploading Balance |
£7110.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16273242
|
Sartorius
MCA8201S-2CEU-0 QP99 |
8200 g | 100 mg | Draft Shield O, QP99 | CEU Electronic Toploading Balance |
£7470.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16273212
|
Sartorius
MCA32202P-2CEU-0 QP99 |
32200 g | 10 mg, 100 mg | Draft Shield O, QP99 | CEU Electronic Toploading Balance |
£15420.00
1 unit |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
16415945
|
Sartorius
MCA5203S-2CEU-R QP99 |
5200 g | 1 mg | Draft Shield R,QP99 | CEU Electronic Toploading Balance |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
16436525
|
Sartorius
MCA6202S-2CEU-0 QP99 |
6200 g | 10 mg | Draft Shield O | CEU Electronic Toploading Balance |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
The New Generation of Modular Premium Laboratory Balances- Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry.
- One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series.
- All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems.
- These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement.
- It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifications
Balance with Type Approval Certificate for China | |
Color Touchscreen | |
100 – 240 VAC (±10%), 50 - 60 Hz, 1.0 A |
D-sub connector, 9-pin | |
Aluminum, plastic PBT, Optiwhite float glass and stainless steel |