Learn More
Sartorius Cubis™ II Semi-Micro Lab Balance
Offers readability of 10 μg and a maximum weighing capacity up to 220 g. Used for weighing small sample amounts down to the μg-range.
Brand:  Sartorius MCA225P-2S00-I QP99
Additional Details : Weight : 6.40000kg
 View more versions of this product
Product Code. 16374508
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifications
Specifications
Balance with Type Approval Certificate for China
60 g, 120 g, 220 g
0.01 mg, 0.02 mg, 0.05 mg
Aluminum, Plastic PBT, Optiwhite Float Glass, and Stainless Steel
2 sec.
100/240 V (± 10%), 50/60 Hz, 1.0 A
D-sub connector, 9-pin
Color Touchscreen
Internal Calibration
Balance with Draft Shield I
Analytical Balance