Learn More
Sartorius Cubis™ II Semi-Micro Lab Balance
Offers readability of 10 μg and a maximum weighing capacity up to 220 g. Used for weighing small sample amounts down to the μg-range.
Brand:  Sartorius MCA125S-2S00-A QP99
Additional Details : Weight : 13.60000kg
 View more versions of this product
Product Code. 16463195
  • £14990.00 / Each
Estimated Shipment: 01-10-2024
to see stock.
Description
Description
The New Generation of Modular Premium Laboratory Balances
Weighing in a regulated environment can be complex, with stringent compliance directives and controls required to deliver the accuracy and reliability of results required by the pharmaceutical industry. One of the technologies helping scientists to achieve this level of accuracy is the Cubis™ II balance series from Sartorius.
All weighing modules of the Cubis™ II balance are designed for intuitive operation, further aided by intelligent diagnostic systems. These design elements guarantee a higher degree of repeatability for different workflows while lowering the probability of human error during measurement. It offers modern user interfaces, pharmaceutical and GxP compliance, including data handling, data integrity and connectivity, ergonomic sample handling, easy process integration, and unlimited communication at the highest level of accuracy and precision.
Specifications
Specifications
Balance with Type Approval Certificate for China
120 g
0.01 mg
Aluminum, Plastic PBT, Optiwhite Float Glass, and Stainless Steel
2 sec.
100/240 V (± 10%), 50/60 Hz, 1.0 A
D-sub connector, 9-pin
Color Touchscreen
Internal Calibration
Balance with Draft Shield A
Analytical Balance
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
Certificats