Learn More
Cumene, 99.9%, extra pure, Thermo Scientific Chemicals
£14.27 - £59.40
Chemical Identifiers
CAS | 98-82-8 |
---|---|
Molecular Formula | C9H12 |
Molecular Weight (g/mol) | 120.19 |
MDL Number | MFCD00008881 |
InChI Key | RWGFKTVRMDUZSP-UHFFFAOYSA-N |
Synonym | isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano |
PubChem CID | 7406 |
ChEBI | CHEBI:34656 |
IUPAC Name | cumene |
SMILES | CC(C)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10748611
|
Thermo Scientific Acros
329730050 |
5 mL | Glass bottle |
£14.27
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10653132
|
Thermo Scientific
329735000 |
500 mL | Glass bottle | N/A | |||||
13468518
|
Thermo Scientific Acros
329730025 |
2.5 L | Glass Bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
98-82-8 | |
120.19 | |
RWGFKTVRMDUZSP-UHFFFAOYSA-N | |
7406 | |
cumene |
C9H12 | |
MFCD00008881 | |
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano | |
CHEBI:34656 | |
CC(C)C1=CC=CC=C1 |
Specifications
98-82-8 | |
Colorless | |
152.0°C to 154.0°C | |
Authentic | |
C9H12 | |
1.4905 to 1.4925 | |
05,393 | |
0.862 | |
Solubility in water: 0.05g/L (20°C). Other solubilities: soluble in alcohol and many other organic solvents,carbon tetrachloride,ether,benzene,acetone | |
CC(C)C1=CC=CC=C1 | |
120.19 | |
CHEBI:34656 | |
120.19 | |
Extra Pure | |
Cumene, 99.9% |
-96.0°C | |
0.8620g/mL | |
31°C | |
99.85% min. (GC) | |
C6H5CH(CH3)2 | |
MFCD00008881 | |
15,2607 | |
isopropylbenzene, 2-phenylpropane, cumol, benzene, 1-methylethyl, 1-methylethyl benzene, isopropylbenzol, cumeen, isopropilbenzene, isopropylbenzeen, 2-fenilpropano | |
RWGFKTVRMDUZSP-UHFFFAOYSA-N | |
cumene | |
7406 | |
0.79 mPa.s (20°C) | |
99.9% | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortabl
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-704-5
missing translation for 'rtecsNumber' : GR8575000
missing translation for 'tsca' : TSCA
- Cumene
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P331-Do NOT induce vomiting.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.