Learn More
Micronova™ CurtainCleaner™ Irradiated Slipcovers
GREENER_CHOICEBIOPHARMA
These irradiated, 100% knit polyester slipcovers work with the Curtain Cleaner adapter to provide a simple method to sanitize cleanroom strip curtains by clamping the top of the strip and sliding down, effectively cleaning both sides at once.
£444.00 - £787.00
Specifications
Product Type Irradiated Mop Slipcover
Color White
Sterility Sterile
Autoclavable Autoclavable
Resistant To Chemicals
View More Specs
Products 2
Product Code Brand Material Dimensions Price Quantity & Availability  
Product Code Brand Material Dimensions Price Quantity & Availability  
11922465
GREENER_CHOICEBIOPHARMA
View Documents
Micronova™
CCSC-6IR
Smooth Knit Polyester, Urethane Foam 30.5 x 5.1 cm (12 x 2 in.)
£444.00
Pack of 12
Estimated Shipment: 08-07-2025
to see stock.
Add to basket
12775705
GREENER_CHOICEBIOPHARMA
View Documents Promotion Details
Micronova™
CCSC-612IR
Smooth Knit Polyester, Urethane Foam 36.2 x 6.4 cm (14.25 x 2.5 in.)
£787.00
12 pairs
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
Description
Description
The Micronova Irradiated CurtainCleaner™ SlipCovers work with the CurtainCleaner Adapter/Handle (CCH-48) to provide a simple and effective method to clean strip curtains in the cleanroom, without the need for a ladder or stool. The covers slide onto the dual 12-inch bars that clamp onto the curtain at the top and slide down to apply or “squeegee” the cleaning agent on both sides of the curtain at once. Covers are available in smooth 100% knit polyester, with two thickness options. Gamma irradiated covers are ideal for aseptic areas.
  • Slipcovers are 100% knit polyester over thick foam
  • Cleans both sides of strip curtains at once
  • Excellent for cleaning and sanitizing
  • Covers fit over two 12-inch bar clamps
  • Gamma irradiated for aseptic areas
  • Launderable for re-use
  • Autoclavable
Order Info
Handle sold separately (Cat. No. 10-041-766A).
Specifications
Specifications
Irradiated Mop Slipcover
Sterile
Chemicals
Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
Smooth Knit Polyester, Urethane Foam
White
Autoclavable
Reusable
Cleaning and Sanitization Cleanroom Strip Curtains
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Micronova™ CurtainCleaner™ Irradiated Slipcovers