Learn More
Ambion™ Custom mirVana™ miRNA Inhibitor
Custom mirVana™ miRNA Inhibitor
Brand:  Ambion™ 4464086
 View more versions of this product
Product Code. 15551511
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically inhibit endogenous miRNAs for the purpose of functional analysis via down-regulation. Use our GeneAssist™ miRNA Workflow Builder to design mimics and inhibitors for any miRNA sequence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Room temperature
mirVana Custom miRNA Inhibitor, 5nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
mirVana Custom miRNA Inhibitor
5 nmol