Learn More
Ambion™ Custom mirVana™ miRNA Mimic
Custom mirVana™ miRNA Mimic
Brand: Ambion™ 4464069
551.76 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. Use our GeneAssist™ miRNA Workflow Builder to design both mimics and inhibitors for any miRNA sequence.Specifications
Room temperature | |
Customer-defined mirVana™ miRNA mimic, 20nmol, miRNA mimics are small, Chemically Modified Double-Stranded RNAs that mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-Regulation of miRNA Activity | |
Customer-defined mirVana™ miRNA mimic | |
20 nmol |
For Research Use Only. Not for use in diagnostic procedures.