missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Custom PreAmp Pools
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design.
£286.00
Specifications
Content And Storage | Pool of assay primers, -20 C |
---|---|
Green Features | Sustainable packaging |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15594354
|
Applied Biosystems™
4478258 |
CUSTOM MIRNA PREAMP POOL (192 A) |
£286.00
1mL |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||||
Description
Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.
Specifications
Pool of assay primers, -20 C | |
Sustainable packaging |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title