missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUX1/Protein CASP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA598395
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
CUX1/Protein CASP | |
Polyclonal | |
Unconjugated | |
CUX1 | |
CASP, CCAAT displacement protein, CDP, CDP/Cut, CDP1, CDP2, Cdpl1, Clox, COY1, cut homolog, cut like homeobox 1, CUTL1, cut-like 1, cut-like homeobox 1, Cux, Cux/CDP, Cux/CDP homeoprotein, Cux1, Cux-1, golgi integral membrane protein 6, GOLIM6, Homeobox protein cut-like 1, homeobox protein cux-1, Kiaa4047, Nbla10317, p100, p110, p200, p75, protein CASP, putative protein product of Nbla10317 | |
Rabbit | |
Protein G | |
RUO | |
116639, 13047, 1523 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3.7 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300, pH 7.4 | |
P39880, P53565, P70403, Q13948 | |
CUX1 | |
Recombinant Human Protein CASP protein (50-210AA) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CUX1/Protein CASP Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction