missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11733-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
CUX1 | |
Polyclonal | |
Unconjugated | |
CUX1 | |
CASP, CCAAT displacement protein, CDP, CDP/Cut, CDP1, Clox, COY1, cut like homeobox 1, CUTL1, CUX, Cux/CDP, CUX1, GOLIM6, Homeobox protein cut like 1, Homeobox protein cux 1, Nbla10317, p100, p110, p200, p75 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116639, 13047, 1523 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53565, P70403, Q13948 | |
CUX1 | |
CUX1 Fusion Protein Ag2335 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |