missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | CUX1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16819013
|
Proteintech
11733-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16809013
|
Proteintech
11733-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
CUX1 | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P53565, P70403, Q13948 | |
116639, 13047, 1523 | |
CUX1 Fusion Protein Ag2335 | |
Primary | |
-20°C | |
CUX1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CASP, CCAAT displacement protein, CDP, CDP/Cut, CDP1, Clox, COY1, cut like homeobox 1, CUTL1, CUX, Cux/CDP, CUX1, GOLIM6, Homeobox protein cut like 1, Homeobox protein cux 1, Nbla10317, p100, p110, p200, p75 | |
CUX1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |