missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CY 09

Inflammasomes Inhibitors

£196.00 - £706.00

Specifications

Chemical Name or Material 4-[[4-Oxo-2-thioxo-3-[[3-(trifluoromethyl)phenyl]methyl]-5-thiazolidinylidene]methyl]benzoic acid
CAS 1073612-91-5
Molecular Formula C19H12F3NO3S2
Purity 98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18783198
View Documents
Tocris Bioscience™
6436/10
10 mg
£196.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18791108
View Documents
Tocris Bioscience™
6436/50
50 mg
£706.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NLRP3 inhibitor (Kd = 500 nM); directly binds to ATP-binding motif of NLRP3 NACHT domain. Blocks NLRP3 inflammasome assembly and activation. Displays therapeutic effects on mouse models of cryopyrin-associated autoinflammatory syndrome (CAPS) and type 2 diabetes. Exhibits no activity for hERG (10 μM).
Specifications

Specifications

4-[[4-Oxo-2-thioxo-3-[[3-(trifluoromethyl)phenyl]methyl]-5-thiazolidinylidene]methyl]benzoic acid
1073612-91-5
C19H12F3NO3S2
98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.