missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cy3 NHS Ester Mono-Reactive CyDye

Use for highly accurate labeling of amine groups. Cytiva Cy3 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

Brand:  Cytiva PA13104

 View more versions of this product

Product Code. 11545045

  • £1609.00 / 10mg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

CyDye fluors are versatile fluorophores designed for a broad range of applications. CyDye fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye flours enables multiplexing without crosstalk for high accuracy.

Cy3 NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dried
10 mg
Foil packs of pre-measured dried dye
Soluble in aqueous solutions
Cy3
For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP)
Cy3 NHS Ester
3 to 10
2°C to 8°C
Red
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Certificates
Special Offers

Special Offers