missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cy3 NHS Ester Mono-Reactive CyDye

Use for highly accurate labeling of amine groups. Cytiva Cy3 NHS Ester Mono-Reactive CyDye contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

£308.00 - £5560.00

Specifications

Format Dried
Product Type Cy3 NHS Ester
Kit Contents Foil packs of pre-measured dried dye
pH Range 3 to 10
Label or Dye Cy3
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11595075
View Documents
Cytiva
PA13106
25 mg
£2817.00
25mg
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
11535045
View Documents
Cytiva
PA13102
50 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10524205
View Documents Promotion Details
Cytiva
PA13101
1 mg
£308.00
1mg
In Stock
to see stock.
Add to basket
 
11545045
View Documents
Cytiva
PA13104
10 mg
£1609.00
10mg
In Stock
to see stock.
Add to basket
 
11585075
View Documents
Cytiva
PA13105
5 mg
£1124.00
5mg
In Stock
to see stock.
Add to basket
 
Description

Description

CyDye fluors are versatile fluorophores designed for a broad range of applications. CyDye fluors produce high sensitivity and highly reproducible performance at a biologically suitable pH range of 3 to 10. The use of multicolor CyDye flours enables multiplexing without crosstalk for high accuracy.

Cy3 NHS Ester provides targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides.

  • Readily dissolves in and produces a stable signal in aqueous buffers (no need for organic solvents)
  • High purity and thorough QC testing generate high levels of chromophore and reactive dye content
  • Narrow excitation and emission bands result in discrete signals from each fluor
  • Demonstrates larger polarization range than conventional fluors
  • Fluorescence lifetime of 2.8 ns
  • High fluorescence intensity
  • Low non-specific binding
Specifications

Specifications

Dried
Foil packs of pre-measured dried dye
Cy3
Cy3 NHS Ester
3 to 10
Red
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.