Learn More
Epredia™ Lab Vision™ Cyclin D1/Bcl-1, Rabbit Monoclonal Antibody
Obtain accurate and reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Cyclin D1/Bcl-1, Rabbit Monoclonal Antibody.
Brand: Epredia™ RM-9104-S0
204.12 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
Description
Cyclin D1 or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).Host Species: Rabbit
Clone: SP4
Isotype: IgG
Species Reactivity: Human, Mouse, Rat. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide from C-terminus of human cyclin D1
Molecular Weight: 36 kDa
Positive Control: MAD109 cells, breast CA or mantle cell lymphoma
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Cyclin D1/Bcl-1 | |
Monoclonal | |
Unconjugated | |
A synthetic peptide from C-terminus of human cyclin D1 | |
IVD | |
Primary | |
Concentrated |
Western Blot | |
SP4 | |
Rabbit | |
100 μL | |
Cell Cycle | |
Human, Mouse | |
IgG |
Safety and Handling
- APD Primary Antibody Products, Antibody Diluent
Signal Word
- Danger
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P363-Wash contaminated clothing before reuse.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.