Learn More
Cyclobutyl bromide, 97%, pure, Thermo Scientific Chemicals
£54.90 - £795.00
Chemical Identifiers
CAS | 4399-47-7 |
---|---|
Molecular Formula | C4H7Br |
Molecular Weight (g/mol) | 135.01 |
MDL Number | MFCD00001317 |
InChI Key | KXVUSQIDCZRUKF-UHFFFAOYSA-N |
Synonym | cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h |
PubChem CID | 78110 |
IUPAC Name | bromocyclobutane |
SMILES | C1CC(C1)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521161
|
Thermo Scientific Acros
320500010 |
1 g | Glass bottle |
£54.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10794104
|
Thermo Scientific Acros
320500050 |
5 g | Glass bottle |
£177.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15199263
|
Thermo Scientific Acros
320500250 |
25 g | Glass Bottle |
£795.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4399-47-7 | |
135.01 | |
KXVUSQIDCZRUKF-UHFFFAOYSA-N | |
78110 | |
C1CC(C1)Br |
C4H7Br | |
MFCD00001317 | |
cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h | |
bromocyclobutane |
Specifications
4399-47-7 | |
1.4300g/mL | |
22°C | |
96% min. (GC) | |
C4H7Br | |
MFCD00001317 | |
cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h | |
KXVUSQIDCZRUKF-UHFFFAOYSA-N | |
bromocyclobutane | |
78110 | |
97% | |
Liquid |
Colorless | |
108.0°C | |
Authentic | |
C4H7Br | |
1.4776 to 1.4797 | |
1.43 | |
Solubility in water: immiscible | |
C1CC(C1)Br | |
135.01 | |
135.01 | |
Pure | |
Cyclobutyl bromide |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a g
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 224-530-9
RUO – Research Use Only