missing translation for 'onlineSavingsMsg'
Learn More
Cyclohexanecarbonyl chloride, 97+%, Thermo Scientific Chemicals Product Code.: 11438677

Cyclohexanecarbonyl chloride, 97+%, Thermo Scientific Chemicals

Product Code. 11438677
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11438677

Brand: Thermo Scientific Alfa Aesar A19824.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2719-27-9
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00001456
InChI Key RVOJTCZRIKWHDX-UHFFFAOYSA-N
Synonym hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chlorideShow MoreShow Less
PubChem CID 75938
IUPAC Name cyclohexanecarbonyl chloride
SMILES ClC(=O)C1CCCCC1

Specifications

Density 1.088
Boiling Point 183°C to 185°C
Flash Point 66°C (150°F)
Refractive Index 1.47
Quantity 25 g
UN Number UN3265
Beilstein 742163
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 146.62
Percent Purity ≥97%
Chemical Name or Material Cyclohexanecarbonyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclohexanecarbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cyclohexanecarbonylchloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexanecarbonyl chloride, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.