Learn More
Cyclohexanecarbonyl chloride, 97+%, Thermo Scientific Chemicals
£19.90 - £49.45
Chemical Identifiers
CAS | 2719-27-9 |
---|---|
Molecular Formula | C7H11ClO |
Molecular Weight (g/mol) | 146.61 |
MDL Number | MFCD00001456 |
InChI Key | RVOJTCZRIKWHDX-UHFFFAOYSA-N |
Synonym | hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride |
PubChem CID | 75938 |
IUPAC Name | cyclohexanecarbonyl chloride |
SMILES | ClC(=O)C1CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11438677
|
Thermo Scientific Alfa Aesar
A19824.14 |
25 g |
£19.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448677
|
Thermo Scientific Alfa Aesar
A19824.22 |
100 g |
£49.45
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.
Solubility
Reacts with water.
Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
2719-27-9 | |
146.61 | |
RVOJTCZRIKWHDX-UHFFFAOYSA-N | |
75938 | |
ClC(=O)C1CCCCC1 |
C7H11ClO | |
MFCD00001456 | |
hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride | |
cyclohexanecarbonyl chloride |
Specifications
2719-27-9 | |
183°C to 185°C | |
C7H11ClO | |
MFCD00001456 | |
742163 | |
hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride | |
RVOJTCZRIKWHDX-UHFFFAOYSA-N | |
cyclohexanecarbonyl chloride | |
75938 | |
≥97% |
1.088 | |
66°C (150°F) | |
1.47 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
ClC(=O)C1CCCCC1 | |
146.61 | |
146.62 | |
Cyclohexanecarbonyl chloride |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 220-322-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Cyclohexanecarbonyl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Cyclohexanecarbonylchloride
RUO – Research Use Only