missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanol, 99%

Product Code. 11438207
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11438207

Brand: Thermo Scientific Alfa Aesar A17576.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 108-93-0 | C6H12O | 100.161 g/mol

Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.

Solubility
Miscible with ethanol, ethyl ether acetone, chloroform, ethyl acetate, linseed oil and benzene. Slightly miscible with cold water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, hydrogen peroxide and nitric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-93-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00003855
InChI Key HPXRVTGHNJAIIH-UHFFFAOYSA-N
Synonym cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol
PubChem CID 7966
ChEBI CHEBI:18099
IUPAC Name cyclohexanol
SMILES C1CCC(CC1)O

Specifications

Melting Point 23°C to 24°C
Density 0.951
Boiling Point 160°C to 161°C
Flash Point 67°C (152°F)
Assay Percent Range 99%
Refractive Index 1.465
Quantity 500 mL
Beilstein 906744
Merck Index 14,2725
Solubility Information Miscible with ethanol,ethyl ether acetone,chloroform,ethyl acetate,linseed oil and benzene. Slightly miscible with cold water.
Formula Weight 100.16
Percent Purity 99%
Chemical Name or Material Cyclohexanol
Show More Show Less
compliance-icons
Product Identifier
  • Cyclohexanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Cyclohexanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.