Learn More
Cyclohexanol, 99%, Thermo Scientific Chemicals
£19.70 - £61.51
Chemical Identifiers
CAS | 108-93-0 |
---|---|
Molecular Formula | C6H12O |
Molecular Weight (g/mol) | 100.161 |
MDL Number | MFCD00003855 |
InChI Key | HPXRVTGHNJAIIH-UHFFFAOYSA-N |
Synonym | cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol |
PubChem CID | 7966 |
ChEBI | CHEBI:18099 |
IUPAC Name | cyclohexanol |
SMILES | C1CCC(CC1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11428207
|
Thermo Scientific Alfa Aesar
A17576.AE |
100 mL |
£19.70
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438207
|
Thermo Scientific Alfa Aesar
A17576.AP |
500 mL |
£21.81
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448207
|
Thermo Scientific Alfa Aesar
A17576.0F |
2500 mL |
£61.51
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclohexanol is used in making soaps, dry cleaning agents and plasticizers. It acts as a solvent for rubber, resins, nitrocellulose, metallic soaps, oils, esters and ethers. It is used in the preparation of adipic acid, hexamethylene diamine, cyclohexanone and caprolactam. It acts as a soap stabilizing agent, disinfection medicated soap and detergent emulsion. It is also useful for paint blending agent, leather degreasing agent and polishes.
Solubility
Miscible with ethanol, ethyl ether acetone, chloroform, ethyl acetate, linseed oil and benzene. Slightly miscible with cold water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, hydrogen peroxide and nitric acid.
Chemical Identifiers
108-93-0 | |
100.161 | |
HPXRVTGHNJAIIH-UHFFFAOYSA-N | |
7966 | |
cyclohexanol |
C6H12O | |
MFCD00003855 | |
cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol | |
CHEBI:18099 | |
C1CCC(CC1)O |
Specifications
108-93-0 | |
0.951 | |
67°C (152°F) | |
C6H12O | |
MFCD00003855 | |
14,2725 | |
Miscible with ethanol,ethyl ether acetone,chloroform,ethyl acetate,linseed oil and benzene. Slightly miscible with cold water. | |
C1CCC(CC1)O | |
100.161 | |
CHEBI:18099 | |
99% |
23°C to 24°C | |
160°C to 161°C | |
99% | |
1.465 | |
906744 | |
cyclohexyl alcohol, hexahydrophenol, hydrophenol, hydroxycyclohexane, hexalin, 1-cyclohexanol, hydralin, adronal, naxol, adronol | |
HPXRVTGHNJAIIH-UHFFFAOYSA-N | |
cyclohexanol | |
7966 | |
100.16 | |
Cyclohexanol |
Safety and Handling
GHS H Statement
H302-H332-H315-H335-H227
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H332-H315-H319-H335
missing translation for 'einecsNumber' : 203-630-6
missing translation for 'rtecsNumber' : GV7875000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : Hygroscopic
Recommended Storage : Ambient temperatures
- Cyclohexanol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Cyclohexanol
RUO – Research Use Only