missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Cycloheximide, 90% Product Code.: 15478109

Thermo Scientific Chemicals Cycloheximide, 90%

Product Code. 15478109
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15478109

Brand: Thermo Scientific Chemicals J66665.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15H23NO4
Molecular Weight (g/mol) 281.35
MDL Number MFCD00082346
InChI Key YPHMISFOHDHNIV-QTEFRXOUNA-N
Synonym 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide; Actidione
PubChem CID 6197
ChEBI CHEBI:27641
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1

Specifications

Type Cycloheximide
Physical Form Powder
Melting Point 111°C to 116°C
Color Cream to White
Quantity 1 g
Flash Point >110°C (230°F)
UN Number UN2811
Beilstein 88868
Boiling Point 492°C
Merck Index 14,2728
Solubility Information Soluble in water at 20mg/mL
Formula Weight 281.35
Percent Purity 90%
Chemical Name or Material Cycloheximide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN18L006 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberJ66665.06,J66665.03
Lot NumberT23K013 Certificate TypeCertificate of Analysis Date02/08/2024 Catalog NumberJ66665.06,J66665.03
Lot NumberP14K029 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog NumberJ66665.06,J66665.03
Lot NumberM23K032 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog NumberJ66665.06,J66665.03
Lot NumberZ12J019 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog NumberJ66665.06,J66665.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Cycloheximide, 90% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.