Learn More
Thermo Scientific Chemicals Cycloheximide, 95%
Cycloheximide, CAS # 66-81-9, is a compound produced by the bacterium Streptomyces griseus that shows an antifungal antibiotic activity with the ability to inhibit protein synthesis only in eukaryotic cells.
£72.06 - £788.93
Chemical Identifiers
CAS | 66-81-9 |
---|---|
Molecular Formula | C15H23NO4 |
Molecular Weight (g/mol) | 281.35 |
MDL Number | MFCD00082346 |
InChI Key | YPHMISFOHDHNIV-QTEFRXOUNA-N |
Synonym | cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide |
PubChem CID | 6197 |
ChEBI | CHEBI:27641 |
IUPAC Name | 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione |
SMILES | [H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10286291
|
Thermo Scientific Chemicals
357420010 |
1 g |
£72.06
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10159861
|
Thermo Scientific Chemicals
357420050 |
5 g |
£236.18
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15214714
|
Thermo Scientific Chemicals
357420250 |
25 g |
£788.93
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Cycloheximide is a semisynthetic compound showing an antifungal antibiotic activity.
- It is produced by the bacterium Streptomyces griseus
- Cycloheximide can inhibit protein synthesis in eukaryotic cells but not in prokaryotes by interacting directly with the translocase enzyme and interfering with the translocation step. Furthermore, this compound also blocks the A site of ribosomes
Application
- Cycloheximide shows a neuroprotective, fungicide, and anticoronaviral activities
- In biomedical research, it can induce glycogenolysis, gluconeogenesis, and ureagenesis
- Due to significant toxic side effects such as teratogenesis, DNA damage, birth defects, and toxicity to sperm, this chemical compound is only used in in vitro research applications
- This compound also shows inhibitory activity against ferroptosis
- It is also used in laboratory media as a selective agent to permit the isolation of pathogenic and non-pathogenic fungi from other types of specimens
- In laboratory research related to plant biology, it can induce ethylene production to promote the plant growth
- In laboratory research, this compound can be applied to determine the half-life of protein in any cellular system. The treatment of in vitro cells with cycloheximide in a time-course experiment followed by western blotting using this cell lysate can show differences in protein half-life.
Chemical Identifiers
66-81-9 | |
281.35 | |
YPHMISFOHDHNIV-QTEFRXOUNA-N | |
6197 | |
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione |
C15H23NO4 | |
MFCD00082346 | |
cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide | |
CHEBI:27641 | |
[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1 |
Specifications
66-81-9 | |
White | |
94% min. (HPLC) | |
C15H23NO4 | |
15,2721 | |
− 28.50 | |
Solubility in water: 2.1 g/100mL (2 c). Other solubilities: soluble in chloroform, ether, acetone, methanol, and ethanol | |
[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1 | |
281.35 | |
CHEBI:27641 | |
95% | |
Cycloheximide |
107.0°C to 114.0°C | |
Authentic | |
Glass bottle | |
MFCD00082346 | |
− 28.50 (20.00°C c=1,CHCl3) | |
cycloheximide, actidione, cycloheximide, naramycin a, kaken, actidion, actidone, hizarocin, naramycin, neocycloheximide | |
YPHMISFOHDHNIV-QTEFRXOUNA-N | |
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione | |
6197 | |
281.35 | |
Glassy Irregular Shaped Granules |
Safety and Handling
GHS H Statement:
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Suspected of causing genetic defects.
May damage the unborn child.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-636-
- Cycloheximide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H360D-May damage the unborn child.
- H341-Suspected of causing genetic defects.
- H411-Toxic to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.