missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylboronic acid, 95%, Thermo Scientific™

£37.08 - £202.66

Chemical Identifiers

CAS 63076-51-7
Molecular Formula C5H11BO2
Molecular Weight (g/mol) 113.95
InChI Key VTTDFSNKIMAQTB-UHFFFAOYSA-N
Synonym cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
PubChem CID 2734327
IUPAC Name cyclopentylboronic acid
SMILES B(C1CCCC1)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11925831
SDS View Documents
Thermo Scientific Acros
359060010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11935831
SDS View Documents
Thermo Scientific Acros
359060100
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopentylboronic acid, 95%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.