Learn More
Cyclopentylmagnesium chloride, 1M in MeTHF, Thermo Scientific™
£45.01 - £152.00
Chemical Identifiers
CAS | 32916-51-1 |
---|---|
Molecular Formula | C5H9ClMg |
Molecular Weight (g/mol) | 128.88 |
MDL Number | MFCD00001357 |
InChI Key | GROGEABZCKQTJW-UHFFFAOYSA-M |
Synonym | cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium |
PubChem CID | 3599845 |
SMILES | Cl[Mg]C1CCCC1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
32916-51-1 | |
128.88 | |
GROGEABZCKQTJW-UHFFFAOYSA-M | |
3599845 |
C5H9ClMg | |
MFCD00001357 | |
cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium | |
Cl[Mg]C1CCCC1 |
Specifications
32916-51-1 | |
MFCD00001357 | |
Air and moisture sensitive | |
GROGEABZCKQTJW-UHFFFAOYSA-M | |
magnesium(2+) cyclopentanide chloride | |
3599845 | |
1M in MeTHF |
C5H9ClMg | |
UN2924 | |
cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium | |
Cl[Mg]C1CCCC1 | |
128.88 | |
128.88 | |
Cyclopentylmagnesium chloride |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H261-H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures