missing translation for 'onlineSavingsMsg'
Learn More
Cyclopropanemethylamine hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 11378066

Cyclopropanemethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11378066
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11378066

Brand: Thermo Scientific Alfa Aesar L13353.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7252-53-1
Molecular Formula C4H9ClN
Molecular Weight (g/mol) 106.57
MDL Number MFCD00012544
InChI Key MQCZBGGAZQXTLH-UHFFFAOYSA-N
Synonym cyclopropanemethylamine hydrochloride, cyclopropylmethanamine hydrochloride, cyclopropylmethyl ammonium chloride, cyclopropylmethylamine hydrochloride, aminomethylcyclopropane hydrochloride, 1-cyclopropylmethanamine hydrochloride, aminomethyl cyclopropane hydrochloride, cyclopropylmethanaminium chloride, cyclopropylmethylamin hydrochloride, cyclopropylmethanamine-hydrochlorideShow MoreShow Less
PubChem CID 81671
IUPAC Name cyclopropylmethanamine;hydrochloride
SMILES [Cl].NCC1CC1

Specifications

Melting Point 202°C to 205°C
Quantity 1 g
Beilstein 3906936
Sensitivity Hygroscopic
Formula Weight 107.58
Percent Purity 98%
Chemical Name or Material Cyclopropanemethylamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclopropanemethylamine hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.