missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CyDye™ DIGE Fluor Minimal Dye Labeling Kits

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel. Amersham CyDye™ DIGE Fluor Minimal Labeling Kits contain size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.

Brand:  Cytiva 25-8010-65


 View more versions of this product

Product Code. 10279184

  • £2078.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy

All minimal dyes allow the labeling of 50μg protein per labeling reaction. Minimal Dye Labeling Kits contain 2nmol or 5nmol of each of the minimal dyes.

For general 2-D applications where sufficient amounts of sample are available.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Solid
5 nmol of each: Cy2, Cy3, and Cy5
5nM of CyDye DIGE Fluor minimal labeling kit (Cy2, Cy3 and Cy5)
Blue, Green, Red
For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2D electrophoresis gel
Amersham CyDye DIGE Fluor Minimal Labeling Kit
-15°C to -30°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger||

Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P271-Use only outdoors or in a well-ventilated area.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash hands thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P305-IF IN EYES:Immediately call a POISON CENTER/doctor..
  • P330-Rinse mouth.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P363-Wash contaminated clothing before reuse.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.