missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CyDye™ DIGE Fluor Minimal Dye Labeling Kits

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel. Amersham CyDye™ DIGE Fluor Minimal Labeling Kits contain size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.

£832.00 - £2078.00

Specifications

Format Solid
Content And Storage -15°C to -30°C
Color Blue, Green, Red
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10066084
View Documents
Cytiva
28-9345-30
2 nmol of each: Cy2, Cy3, and Cy5
£832.00
1 set
In Stock
to see stock.
Add to basket
 
10279184
View Documents
Cytiva
25-8010-65
5 nmol of each: Cy2, Cy3, and Cy5
£2078.00
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
Description

Description

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy

All minimal dyes allow the labeling of 50μg protein per labeling reaction. Minimal Dye Labeling Kits contain 2nmol or 5nmol of each of the minimal dyes.

For general 2-D applications where sufficient amounts of sample are available.

Specifications

Specifications

Solid
-15°C to -30°C
Blue, Green, Red
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Cy2, Cy3, Cy5

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.