Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 04352814
Additional Details : Weight : 0.10000kg
Description
- With NS socket and cone
- With scale (approx. volume/mL)
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
50 mL | |
29/32 NS | |
Yes |
Borosilicate Glass | |
Pressure Equalizing Tube | |
Dropping Funnel |