Learn More
Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels, with PTFE Plug
Useful for transferring fluids and handling air-sensitive reagents in a sealed, inert-gas environment. Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 04177014
 View more versions of this product
Product Code. 10047810
  • £236.00 / Pack of 2
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With scale (approx. volume/mL)
  • With screwthread retaining device
  • Cylindrical shape with NS socket and cone
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
1000 mL
Without Pressure Equalizing Tube
29/32 NS
Borosilicate Glass
Yes
Dropping Funnel