Learn More
Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels, with PTFE Plug
Useful for transferring fluids and handling air-sensitive reagents in a sealed, inert-gas environment. Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 04175814
Additional Details : Weight : 0.11100kg
Description
- With scale (approx. volume/mL)
- Without pressure equalizing tube
- With screwthread retaining device
- Cylindrical shape with NS socket and cone
- Corrosion-free and resistant to thermal shock
Specifications
500 mL | |
Without Pressure Equalizing Tube | |
29/32 NS |
Borosilicate Glass | |
Yes | |
Dropping Funnel |