missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Cylindrical Reaction Vessel, Flat Bottom

Brand:  Lenz Laborglasinstrumente™ 06000482

Code : 50

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11522174

  • £561.80 / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

DURAN™ Tubing
Borosilicate Glass, Duran Glass
Laboratory Flange 150
Cylindrical
160 mm
5 L
350 mm
Reaction vessel
Flat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers