Learn More
Lenz Laborglasinstrumente™ Cylindrical Reaction Vessel with Thermostatic Jacket and Withdrawal Valve, Threaded
Cylindrical shape, with graduations
Brand:  Lenz Laborglasinstrumente™ 06030279
 View more versions of this product
Product Code. 11552284
  • £1580.00 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ tubing, cylindrical shape with graduation, laboratory flange LF and withdrawal valve. Thermostatic jacket with either GL 14, GL 18, KF 15 or KF 25 connectors.
Specifications
Specifications
DURAN™ Tubing
Borosilicate Glass, Duran Glass
Laboratory Flange 100
Cylindrical
4 L
495 mm
Reaction vessel