missing translation for 'onlineSavingsMsg'
Learn More

GeneBLAzer™ CysLT2-NFAT-bla CHO-K1 Cells

GeneBLAzer™ CysLT2-NFAT-bla CHO-K1 Cells contain the human Cysteinyl receptor (CysLT2).

Brand:  GeneBLAzer™ K1721

Product Code. 10349103

  • £10780.00 / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gene Symbol: CysLT2
  • Accession Number: NM_020377.2
  • Population Status: Single Cell Clone Origin
  • GeneBLAzer™ CysLT2-NFAT-bla CHO-K1 cells contain the human Cysteinyl receptor (CysLT2) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. no. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
  • The GeneBLAzer™ CysLT2-NFAT-bla CHO-K1 cells are functionally validated for Z' and EC50concentrations of LTD4. In addition, GeneBLAzer™ CysLT2-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
  • Cysteinyl leukotrienes play a role in the patho-physiology of numerous inflammatory disorders, including chronic asthmatic inflammation. The cysteine leukotriene receptors, CysLT1 and CysLT2, are thus primary targets for anti-asthma drugs. A number of cysteinyl leukotrienes (including LTC4, LTD4, and LTE4) have been identified, and are known to function as inflammatory mediators as well as being potent smooth muscle contractile agents.
  • Leukotriene receptor antagonists (LTRAs) reduce the effect of allergic inflammation and athsma by blocking cysteine leukotriene activity. Several CysLT antagonists, such as montelukast, zafirlucast, and pranlukast, are currently used in the treatment of asthma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid Nitrogen
GPCRs
2 x 106 Cells
Fluorescence
CysLT2-NFAT-bla CHO-K1 cells, CysLT2-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Dividing Cells
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers