missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cystatin SA Antibody Pair [HRP]
Mouse Monoclonal Antibody Pair
Brand: Novus Biologicals™ NBP2-79652-5PLATES
Additional Details : Weight : 0.00970kg
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA.Spécification
2.4 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Cystatin SA. | |
Sandwich ELISA | |
1470 | |
Mouse | |
5 Plates | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 150 pg/mL is recommended. |
2.34-150 pg/mL | |
HRP | |
Antibody Pairs | |
Human | |
cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor | |
CST2 | |
RUO | |
Cystatin SA |